Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170710.1 | NP_001164181.1 | 4 | 388 | ||
NM_001170711.1 | NP_001164182.1 | 4 | 394 | |||
NM_030121.4 | NP_084397.3 | 4 | 384 | |||
GenBank mRNA | AF398969.1 | - | 4 | 311 | ||
AK016001.1 | - | 4 | 442 | |||
AK021239.1 | - | 4 | 325 | |||
AK030168.1 | - | 4 | 327 | |||
AK087299.1 | - | 4 | 346 | |||
AK182854.1 | - | 3 | 144 | |||
AK214224.1 | - | 3 | 144 | |||
BC025106.1 | - | 4 | 308 | 4194829 |