Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uvrag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178635.3 | NP_848750.3 | 3 | 447 | ||
GenBank mRNA | AB020334.1 | - | 2 | 52 | ||
AK030814.1 | - | 3 | 378 | |||
AK043084.1 | - | 3 | 425 | |||
AK044129.1 | - | 3 | 430 | |||
AK078085.1 | - | 3 | 439 | |||
AK089900.1 | - | 3 | 432 | |||
AK154903.1 | - | 3 | 447 | |||
AK165015.1 | - | 3 | 435 | |||
AK165185.1 | - | 3 | 427 | |||
BC034176.1 | - | 3 | 416 | 4954871 |