Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030138.2 | NP_084414.1 | 6, 7 | 786 | ||
GenBank mRNA | AB490692.1 | - | 6, 7 | 539 | ||
AK020591.1 | - | 6, 7 | 787 | |||
AK029343.1 | - | 6, 7 | 711 | |||
AK129041.1 | - | 6, 7 | 763 | |||
AK147319.1 | - | 5, 6 | 658 |