Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NINJ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294345.1 | NP_001281274.1 | 2 | 212 | ||
NM_001294346.1 | NP_001281275.1 | 2 | 319 | |||
NM_016533.5 | NP_057617.2 | 2 | 558 | |||
GenBank mRNA | AK296012.1 | - | 2 | 558 | ||
AK298043.1 | - | 2 | 270 | |||
AK315995.1 | - | 2 | 556 | |||
BC057766.1 | - | 2 | 226 | |||
BI597990.1 | - | 2 | 411 | |||
BM128205.1 | - | 2 | 180 | |||
CB995632.1 | - | 2 | 239 | |||
CD695373.1 | - | 2 | 306 |