Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3j1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144545.4 | NP_653128.2 | 2, 3 | 257 | ||
GenBank mRNA | AK019219.1 | - | 2, 3 | 185 | ||
AK082048.1 | - | 1, 2 | 48 | |||
AK147154.1 | - | 2, 3 | 189 | |||
AK148004.1 | - | 2, 3 | 260 | |||
AK154934.1 | - | 2, 3 | 258 | |||
AK161038.1 | - | 2, 3 | 215 |
Gene Symbol: Eif3j2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256055.1 | NP_001242984.1 | 1 | 260 | ||
GenBank mRNA | BC080788.1 | - | 1 | 214 | 4166214 | |
BC108355.1 | - | 1 | 40 |