Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178113.3 | NP_835214.2 | 12, 13 | 1944 | ||
GenBank mRNA | AK129046.1 | - | 11, 12 | 1470 | ||
AK139081.1 | - | 11, 12 | 1502 | |||
BC048190.1 | - | 12, 13 | 1944 | 6831876 | ||
DQ340802.1 | - | 11, 12 | 1599 |