Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rictor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030168.3 | NP_084444.3 | 31 | 3794 | ||
GenBank mRNA | AK173326.1 | - | 7 | 1299 | ||
AY497009.1 | - | 31 | 3772 | |||
AY540053.1 | - | 31 | 3792 | |||
BC120903.1 | - | 27 | 3433 | 40086992 | ||
BC120904.1 | - | 31 | 3791 | 40086995 |