Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030172.2 | NP_084448.1 | 2, 3 | 264 | ||
GenBank mRNA | AK011507.1 | - | 2, 3 | 264 | ||
AK011836.1 | - | 2, 3 | 246 | |||
AK035051.1 | - | 2, 3 | 244 | |||
AK132473.1 | - | 2, 3 | 326 | |||
BC049109.1 | - | 2, 3 | 226 | 6414718 |