Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mctp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030174.2 | NP_084450.2 | 15 | 1812 | ||
GenBank mRNA | AK013379.1 | - | 2 | 418 | ||
AK047562.1 | - | 15 | 1813 | |||
AK054478.1 | - | 11 | 1185 |