Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enthd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183137.2 | NP_898960.2 | 3, 4 | 272 | ||
GenBank mRNA | AK154660.1 | - | 3, 4 | 237 | ||
AK181460.1 | - | 14 | ||||
AK196363.1 | - | 14 | ||||
AK209567.1 | - | 14 | ||||
BB868415.1 | - | 3, 4 | 273 | |||
BC049110.1 | - | 3, 4 | 226 | |||
BC064313.1 | - | 3, 4 | 226 | |||
BC094318.1 | - | 2, 3 | 761 | |||
BC137580.1 | - | 3, 4 | 241 | 8860599 | ||
BC144724.1 | - | 3, 4 | 241 | 9053272 | ||
BY084429.1 | - | 3, 4 | 242 |