Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271483.1 | NP_001258412.1 | 5 | 620 | ||
NM_001271484.1 | NP_001258413.1 | 5 | 705 | |||
NM_030179.3 | NP_084455.2 | 5 | 608 | |||
NM_175378.1 | NP_780587.1 | 5 | 705 | |||
GenBank mRNA | AK017914.1 | - | 2 | 195 | ||
AK018895.1 | - | 5 | 698 | |||
AK019516.1 | - | 5 | 621 | |||
AK029962.1 | - | 5 | 706 | |||
AK075758.1 | - | 5 | 696 | |||
AK161301.1 | - | 5 | 695 | |||
BC035226.1 | - | 5 | 608 | 4920788 |