Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114361.1 | NP_001107833.1 | 22 | 2617 | ||
NM_001114362.1 | NP_001107834.1 | 20 | 2410 | |||
NM_001286567.1 | NP_001273496.1 | 22 | 2520 | |||
NM_199466.3 | NP_955760.3 | 19 | 2281 | |||
GenBank mRNA | AK019611.1 | - | 3 | 239 | ||
AK077527.1 | - | 14 | 1446 | |||
AK139067.1 | - | 5 | 990 | |||
AK144604.1 | - | 22 | 2618 | |||
AK189201.1 | - | 5 | 369 | |||
BC032258.1 | - | 8 | 765 | |||
BC065178.1 | - | 22 | 2458 | 6827397 | ||
BC066099.1 | - | 19 | 2212 | 30533573 | ||
BC067011.1 | - | 19 | 2230 | 6405184 | ||
BC070427.1 | - | 20 | 2350 | 6413041 | ||
BC079619.1 | - | 19 | 2212 | 30533573 | ||
DQ230326.1 | - | 21 | 2300 |