Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198101.1 | NP_932769.1 | 9 | 789 | ||
GenBank mRNA | AK043883.1 | - | 9 | 788 | ||
AK082337.1 | - | 9 | 786 | |||
AK137177.1 | - | 7 | 782 | |||
AK172438.1 | - | 9 | 777 | |||
AK216290.1 | - | 5 | 319 | |||
BC057037.1 | - | 9 | 789 | 5702855 | ||
BC060628.1 | - | 9 | 764 | 6847400 |