Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198101.1 | NP_932769.1 | 19 | 2215 | ||
GenBank mRNA | AK019874.2 | - | 8 | 965 | ||
AK052221.1 | - | 19 | 2181 | |||
AK082337.1 | - | 19 | 2212 | |||
AK137177.1 | - | 17 | 2208 | |||
AK172438.1 | - | 19 | 2203 | |||
BC057037.1 | - | 19 | 2215 | 5702855 | ||
BC060628.1 | - | 19 | 2190 | 6847400 |