Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wsb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042565.3 | NP_001036030.1 | 4 | 1315 | ||
NM_019653.3 | NP_062627.3 | 9 | 1461 | |||
GenBank mRNA | AF033186.1 | - | 9 | 1158 | ||
AK019206.2 | - | 9 | 1439 | |||
AK048511.1 | - | 4 | 1315 | |||
AK145974.1 | - | 9 | 1440 | |||
AK146752.1 | - | 9 | 1436 | |||
AK150394.1 | - | 9 | 1436 | |||
AK150600.1 | - | 9 | 1436 | |||
AK152545.1 | - | 9 | 1439 | |||
AK159905.1 | - | 9 | 1436 | |||
AK167174.1 | - | 9 | 1438 | |||
AK167394.1 | - | 9 | 1439 | |||
AK167942.1 | - | 9 | 1436 | |||
AK168468.1 | - | 9 | 2859 | |||
AK168565.1 | - | 9 | 2858 | |||
AK169058.1 | - | 9 | 1436 | |||
BC019601.1 | - | 4 | 531 |