Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030208.3 | NP_084484.2 | 8 | 1218 | ||
GenBank mRNA | AK019101.1 | - | 8 | 1209 | ||
AK046957.1 | - | 8 | 1220 | |||
AK132402.1 | - | 8 | 1219 | |||
BC152332.1 | - | 8 | 1213 | 40141614 |