Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crispld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030209.3 | NP_084485.1 | 8, 9 | 1106 | ||
GenBank mRNA | AK019034.1 | - | 8, 9 | 1102 | ||
AK033858.1 | - | 8, 9 | 1070 | |||
AK132171.1 | - | 8, 9 | 1049 | |||
AK149909.1 | - | 8, 9 | 1063 | |||
AK149925.1 | - | 8, 9 | 1063 | |||
AK150180.1 | - | 8, 9 | 1063 | |||
AK154308.1 | - | 8, 9 | 1106 | |||
BC090642.1 | - | 8, 9 | 1100 | 30653388 |