Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322802.1 | NP_001309731.1 | 3 | 1018 | ||
NM_005386.3 | NP_005377.1 | 3 | 1023 | |||
NM_181689.2 | NP_859017.1 | 2 | 942 | |||
GenBank mRNA | AB002392.1 | - | 3 | 936 | ||
BC001768.1 | - | 3 | 967 | 3353944 | ||
U25033.1 | - | 3 | 963 | |||
U25034.1 | - | 2 | 884 |