Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080847.3 | NP_543123.2 | 8 | 1168 | ||
GenBank mRNA | AF403043.1 | - | 8 | 1169 | ||
AK019556.1 | - | 8 | 1170 | |||
AK029759.1 | - | 8 | 1100 | |||
AK079418.1 | - | 8 | 1274 | |||
BC050794.1 | - | 8 | 1117 | 6742480 |