Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030224.1 | NP_084500.1 | 3 | 788 | ||
GenBank mRNA | AK019639.1 | - | 3 | 810 | ||
AK085347.1 | - | 2 | 546 | |||
BC120835.1 | - | 3 | 788 | 40129758 | ||
BC137682.1 | - | 3 | 788 | 8860701 |