Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030225.4 | NP_084501.1 | 12 | 996 | ||
GenBank mRNA | AK019713.2 | - | 993 | |||
AK054053.1 | - | 12 | 950 | |||
AK149664.1 | - | 12 | 972 | |||
AK158877.1 | - | 12 | 997 | |||
AK168570.1 | - | 12 | 972 | |||
AK169943.1 | - | 12 | 950 | |||
AK197945.1 | - | 3 | 199 | |||
AK203473.1 | - | 3 | 199 | |||
AK211646.1 | - | 2 | 168 | |||
AK214016.1 | - | 3 | 253 | |||
AK220022.1 | - | 3 | 199 | |||
BC006702.1 | - | 12 | 933 | 3583936 | ||
BC024066.1 | - | 3 | 383 | |||
CT010197.1 | - | 12 | 932 |