Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133779.2 | NP_598540.2 | 9 | 1266 | ||
GenBank mRNA | AK019717.2 | - | 9 | 1256 | ||
AK044499.1 | - | 9 | 1250 | |||
AK049171.1 | - | 9 | 1260 | |||
AK049990.1 | - | 9 | 1259 | |||
AK090020.1 | - | 9 | 1258 | |||
AK153462.1 | - | 9 | 1252 | |||
AK153464.1 | - | 9 | 1252 | |||
AK155057.1 | - | 9 | 1253 | |||
AK157226.1 | - | 9 | 1246 | |||
AK162573.1 | - | 7 | 1464 | |||
BC016520.1 | - | 10 | 1320 | |||
BC034175.1 | - | 9 | 1242 | 5026752 |