Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146085.1 | NP_001139557.1 | 4 | 1277 | ||
NM_001146086.1 | NP_001139558.1 | 3 | 1200 | |||
NM_030236.3 | NP_084512.2 | 2 | 1090 | |||
GenBank mRNA | AK020018.1 | - | 2 | 1091 | ||
AK077162.1 | - | 1 | 243 | |||
AK171630.1 | - | 7 | 1625 | |||
BC048149.1 | - | 4 | 1311 | 5706344 | ||
BC049155.1 | - | 3 | 1142 |