Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024467.3 | NP_077787.3 | 2 | 829 | ||
GenBank mRNA | AK048624.1 | - | 2 | 690 | ||
AK052274.1 | - | 2 | 664 | |||
AK089263.1 | - | 2 | 679 | |||
BC033441.1 | - | 2 | 655 | |||
BC059823.1 | - | 2 | 829 | 6843792 |