Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170552.1 | NP_001164023.1 | 6 | 999 | ||
NM_030675.3 | NP_109600.2 | 6 | 953 | |||
GenBank mRNA | AF306509.1 | - | 6 | 666 | ||
AF310134.1 | - | 6 | 929 | |||
AK039264.1 | - | 974 | ||||
AK085107.1 | - | 6 | 981 | |||
AK144667.1 | - | 6 | 935 | |||
AY328895.1 | - | 5 | 635 | |||
AY464945.1 | - | 5 | 635 | |||
BC016087.1 | - | 6 | 903 | |||
BC044820.1 | - | 5 | 694 | |||
BC054819.1 | - | 6 | 1044 | |||
BC085252.1 | - | 6 | 759 | |||
BC141042.1 | - | 6 | 797 | 9055654 |