Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023508.6 | NP_075997.1 | 5 | 475 | ||
GenBank mRNA | AK005602.1 | - | 5 | 498 | ||
AK005814.1 | - | 5 | 475 | |||
AK006040.1 | - | 4 | 377 | |||
AK006773.1 | - | 5 | 492 | |||
AK132738.1 | - | 5 | 522 | |||
AK133140.1 | - | 4 | 338 | |||
BC048432.1 | - | 5 | 477 | 6773342 |