Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023508.6 | NP_075997.1 | 4 | 326 | ||
GenBank mRNA | AK005602.1 | - | 4 | 349 | ||
AK005814.1 | - | 4 | 326 | |||
AK006040.1 | - | 3 | 228 | |||
AK006773.1 | - | 4 | 343 | |||
AK132738.1 | - | 4 | 373 | |||
AK133140.1 | - | 3 | 189 | |||
BC048432.1 | - | 4 | 328 | 6773342 |