Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030251.3 | NP_084527.2 | 4 | 357 | ||
GenBank mRNA | AB053477.1 | - | 4 | 288 | ||
AK049100.1 | - | 4 | 273 | |||
AK088437.1 | - | 4 | 322 | |||
AK089474.1 | - | 2 | 721 | |||
AK155432.1 | - | 2 | 1320 | |||
AK155505.1 | - | 2 | 1320 | |||
AK155644.1 | - | 2 | 1320 | |||
AK220208.1 | - | 4 | 214 | |||
BC003234.1 | - | 4 | 301 | 3483893 | ||
BY201769.1 | - | 4 | 315 |