Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286433.1 | NP_001273362.1 | 4 | 683 | ||
NM_001286435.1 | NP_001273364.1 | 5 | 895 | |||
NM_001286436.1 | NP_001273365.1 | 5 | 988 | |||
NM_001286438.1 | NP_001273367.1 | 6 | 963 | |||
NM_001286439.1 | NP_001273368.1 | 6 | 972 | |||
NM_001286440.1 | NP_001273369.1 | 5 | 1101 | |||
NM_005009.2 | NP_005000.1 | 5 | 785 | |||
GenBank mRNA | AF086133.1 | - | 3 | 483 | ||
AK094439.1 | - | 2 | 1644 | |||
BC004880.2 | - | 5 | 768 | 3830205 | ||
BC017067.1 | - | 5 | 785 | 3913604 | ||
Y07604.1 | - | 5 | 765 |