Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zxdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030260.3 | NP_084536.2 | 2 | 1201 | ||
NM_173002.3 | NP_766590.1 | 2 | 1201 | |||
GenBank mRNA | AK030287.1 | - | 2 | 1117 | ||
AK042190.1 | - | 2 | 1128 | |||
AK042392.1 | - | 2 | 1111 | |||
AK044429.1 | - | 2 | 1094 | |||
AK044529.1 | - | 2 | 1125 | |||
AK044948.1 | - | 2 | 1095 | |||
AK047079.1 | - | 2 | 1083 | |||
AK047538.1 | - | 2 | 1117 | |||
AK080307.1 | - | 2 | 1095 | |||
AK081005.1 | - | 2 | 1203 | |||
AK141613.1 | - | 2 | 1078 | |||
AK169517.1 | - | 2 | 1088 | |||
AK185703.1 | - | 1 | 114 | |||
AK218430.1 | - | 1 | 114 | |||
BC003332.1 | - | 2 | 1185 | 3256958 |