Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zxdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030260.3 | NP_084536.2 | 5 | 1481 | ||
NM_173002.3 | NP_766590.1 | 5 | 1481 | |||
GenBank mRNA | AK030287.1 | - | 5 | 1397 | ||
AK042190.1 | - | 5 | 1408 | |||
AK042392.1 | - | 5 | 1391 | |||
AK044429.1 | - | 5 | 1374 | |||
AK044529.1 | - | 5 | 1405 | |||
AK044948.1 | - | 5 | 1375 | |||
AK047079.1 | - | 5 | 1363 | |||
AK047538.1 | - | 5 | 1397 | |||
AK080307.1 | - | 5 | 1375 | |||
AK081005.1 | - | 5 | 1483 | |||
AK169517.1 | - | 5 | 1368 | |||
AK205036.1 | - | 2 | 158 | |||
BC003332.1 | - | 5 | 1465 | 3256958 |