Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199242.1 | NP_001186171.1 | 8 | 885 | ||
NM_001199243.1 | NP_001186172.1 | 7 | 783 | |||
NM_001199244.1 | NP_001186173.1 | 7 | 926 | |||
NM_001199245.1 | NP_001186174.1 | 7 | 779 | |||
NM_030265.3 | NP_084541.3 | 7 | 711 | |||
GenBank mRNA | AF305071.1 | - | 7 | 682 | ||
AF453243.1 | - | 7 | 619 | |||
AK039048.1 | - | 7 | 712 | |||
AK049662.1 | - | 7 | 663 | |||
AK133416.1 | - | 7 | 631 | |||
AK148685.1 | - | 7 | 928 | |||
AK165773.1 | - | 7 | 653 | |||
AY647240.1 | - | 7 | 662 | |||
BC051130.1 | - | 8 | 885 | 6532082 | ||
DQ148501.1 | - | 8 | 885 | |||
DQ148502.1 | - | 6 | 643 | |||
DQ148503.1 | - | 7 | 663 | |||
DQ148504.1 | - | 7 | 779 |