Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019573.3 | NP_062519.2 | 8 | 1001 | ||
GenBank mRNA | AF187014.1 | - | 8 | 952 | ||
AK018507.1 | - | 8 | 931 | |||
AK019911.1 | - | 8 | 959 | |||
AK050541.1 | - | 8 | 953 | |||
AK078528.1 | - | 8 | 954 | |||
BC014716.1 | - | 8 | 957 | 4241066 | ||
EF410109.1 | - | 3 | 619 |