Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030691.1 | NP_109616.1 | 2 | 132 | ||
GenBank mRNA | AJ278575.1 | - | 2 | 132 | ||
AK040569.1 | - | 2 | 112 | |||
AK151417.1 | - | 2 | 121 | |||
AK152186.1 | - | 2 | 121 | |||
AK156382.1 | - | 2 | 70 | |||
AK163058.1 | - | 2 | 112 | |||
BC057951.1 | - | 2 | 129 | 5341788 |