Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024954.1 | NP_001020125.1 | 6 | 813 | ||
GenBank mRNA | AJ300183.1 | - | 6 | 741 | ||
AJ300184.1 | - | 3 | 247 | |||
AK010506.1 | - | 3 | 292 | |||
AK038786.1 | - | 417 | ||||
AK076968.1 | - | 4 | 438 | |||
BC032875.1 | - | 5 | 509 | 1263363 |