Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030558.2 | NP_085035.1 | 8 | 980 | ||
GenBank mRNA | AF231122.1 | - | 8 | 932 | ||
AK131988.1 | - | 8 | 1008 | |||
BC019975.1 | - | 8 | 980 | 4223351 |