Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030559.3 | NP_085036.3 | 16 | 1594 | ||
GenBank mRNA | AB056721.1 | - | 16 | 1552 | ||
AK032558.1 | - | 7 | 555 | |||
AK049872.1 | - | 16 | 1551 | |||
AK162984.1 | - | 13 | 1917 | |||
AK169694.1 | - | 13 | 2133 | |||
BC003944.1 | - | 4 | 466 | 3487257 | ||
BC025626.1 | - | 16 | 1560 | 5320572 |