Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030559.3 | NP_085036.3 | 5 | 531 | ||
GenBank mRNA | AK049872.1 | - | 5 | 488 | ||
AK162984.1 | - | 5 | 498 | |||
AK169694.1 | - | 4 | 594 | |||
BC025626.1 | - | 5 | 497 | 5320572 |