Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC004004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030561.3 | NP_085038.3 | 3 | 430 | ||
GenBank mRNA | AK157037.1 | - | 3 | 247 | ||
AK217020.1 | - | 2 | 220 | |||
BC004004.1 | - | 3 | 267 | 3492768 | ||
BC058575.1 | - | 3 | 239 | 6488611 | ||
CD353313.1 | - | 3 | 382 |