Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030563.2 | NP_085040.2 | 2 | 1359 | ||
GenBank mRNA | AK134934.1 | - | 2 | 1285 | ||
AK147522.1 | - | 2 | 1360 | |||
AK170714.1 | - | 2 | 1093 | |||
AK172981.1 | - | 2 | 1341 | |||
BC004022.1 | - | 1 | 135 | |||
BC089324.1 | - | 1 | 524 |