Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030564.1 | NP_085041.1 | 3, 4 | 741 | ||
GenBank mRNA | AF434815.1 | - | 2, 3 | 636 | ||
AK045696.1 | - | 3, 4 | 737 | |||
AK046274.1 | - | 3, 4 | 723 | |||
AK046381.1 | - | 3, 4 | 737 | |||
AK077221.1 | - | 3, 4 | 738 | |||
AK132330.1 | - | 757 | ||||
AK137606.1 | - | 717 | ||||
AK150976.1 | - | 3, 4 | 735 | |||
AK159753.1 | - | 3, 4 | 723 | |||
AK206112.1 | - | 1, 2 | 267 | |||
BC004042.1 | - | 3, 4 | 741 | 3496314 |