Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020036.4 | NP_064420.2 | 1 | 203 | ||
GenBank mRNA | AK003648.1 | - | 1 | 204 | ||
AK009664.1 | - | 1 | 205 | |||
AK009956.1 | - | 1 | 204 | |||
AY293058.1 | - | 1 | 186 | |||
BC060284.1 | - | 1 | 189 | 30293362 | ||
BC145379.1 | - | 1 | 159 | 9053937 |