Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030697.2 | NP_109622.1 | 5, 6 | 1653 | ||
GenBank mRNA | AK002623.1 | - | 5, 6 | 1654 | ||
AK132011.1 | - | 5, 6 | 1675 | |||
KJ650203.1 | - | 4, 5 | 1584 | |||
KJ663713.1 | - | 4, 5 | 1425 | |||
KJ663714.1 | - | 5, 6 | 1446 |