Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030697.2 | NP_109622.1 | 9 | 2194 | ||
GenBank mRNA | AK002623.1 | - | 9 | 2195 | ||
AK132011.1 | - | 9 | 2216 | |||
BC037741.1 | - | 4 | 328 | |||
BC061055.1 | - | 4 | 340 | |||
BC115933.1 | - | 3 | 298 | |||
KJ650203.1 | - | 8 | 2125 | |||
KJ663713.1 | - | 8 | 1966 | |||
KJ663714.1 | - | 9 | 1987 |