Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163348.1 | NP_001156820.1 | 5 | 2173 | ||
NM_030699.2 | NP_109624.1 | 5 | 2173 | |||
GenBank mRNA | AB038666.1 | - | 4 | 1078 | ||
AB038667.1 | - | 5 | 1950 | |||
AF475069.1 | - | 4 | 1078 | |||
AF475070.1 | - | 4 | 1078 | |||
AF475075.1 | - | 4 | 1078 | |||
AF475076.1 | - | 4 | 1078 | |||
AK048796.1 | - | 4 | 1039 | |||
AK134467.1 | - | 4 | 2003 |