Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030713.2 | NP_109638.2 | 7 | 1270 | ||
GenBank mRNA | AF292648.1 | - | 7 | 1260 | ||
AF292649.1 | - | 7 | 1256 | |||
AK170181.1 | - | 6 | 1141 | |||
AK172368.1 | - | 7 | 1270 | |||
BC049772.1 | - | 7 | 1198 | 6334069 |