Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030719.3 | NP_109644.2 | 8 | 1182 | ||
GenBank mRNA | AJ296173.1 | - | 8 | 1222 | ||
AK039122.1 | - | 8 | 1208 | |||
AK052217.1 | - | 8 | 1167 | |||
BC094613.1 | - | 8 | 1182 | 6856325 |