Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030724.3 | NP_109649.1 | 2, 3 | 519 | ||
GenBank mRNA | AF236636.1 | - | 2, 3 | 524 | ||
AK051894.1 | - | 2, 3 | 289 | |||
AK053745.1 | - | 2, 3 | 479 | |||
AK077517.1 | - | 2, 3 | 456 | |||
AK082837.1 | - | 2, 3 | 482 | |||
AK155939.1 | - | 2, 3 | 483 | |||
AK168318.1 | - | 2, 3 | 497 | |||
BC023789.1 | - | 2, 3 | 483 | 5352192 |