Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020267.2 | NP_064663.2 | 3 | 1183 | ||
GenBank mRNA | AJ249129.1 | - | 3 | 1132 | ||
AJ278191.1 | - | 3 | 1191 | |||
BC039979.1 | - | 3 | 609 | |||
BC045602.1 | - | 3 | 598 | |||
BC098369.1 | - | 3 | 1152 | 6830438 |