Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020267.2 | NP_064663.2 | 3 | 1027 | ||
GenBank mRNA | AJ249129.1 | - | 3 | 976 | ||
AJ278191.1 | - | 3 | 1035 | |||
BC039979.1 | - | 3 | 453 | |||
BC045602.1 | - | 3 | 442 | |||
BC098369.1 | - | 3 | 996 | 6830438 |